Learn More
TCR γδ Hamster anti-Mouse, PE, Clone: GL3, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 553178
Description
The GL3 antibody reacts with a common epitope of the δ chain of the T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with αβ TCR-bearing T cells. In
Host Species: Hamster
Clone: GL3
Isotype: IgG2 κ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Flow Cytometry
Specifications
| TCR γδ | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| GL3 | |
| PE | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.