Learn More
trefoil factor 3 (intestinal), Mouse, Clone: 3G11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant TFF3.
Brand: Abnova H00007033-M02.50ug
Description
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer and affect healing of the epithelium. This gene is expressed in goblet cells of the intestines and colon. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq
Sequence: EEYVGLSANQCAVPAKDRVDCGYPHVTPKECNNRGCCFDSRIPGVPWCFKPLQEAECTFSpecifications
| trefoil factor 3 (intestinal) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC017859 | |
| TFF3 | |
| TFF3 (AAH17859.1, 15 a.a. ∼ 73 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 50 μg | |
| Yes | |
| 7033 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3G11 | |
| Mouse monoclonal antibody raised against a full length recombinant TFF3. | |
| TFF3 | |
| HITF/ITF/TFI/hP1.B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.