missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-β1 Rat anti-Human, Pig, Biotin, Clone: A75-3, BD
Description
The A75-3 antibody reacts with mouse, pig, human TGF-β1. The immunogen used to generate this hybridoma was recombinant mouse TGF-β1. This antibody is routinely tested by ELISA detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
ELISA
Specifications
Specifications
| Antigen | TGF-β1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | A75-3 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Rat |
| Immunogen | Recombinant mouse TGF-β1 |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?