Learn More
TGFB1, Mouse, Clone: 4E1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TGFB1.
Brand: Abnova H00007040-M02A.200uL
Description
TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM
Sequence: ALDTNYCFSSTEKNCCVRQLYIDFRKDLGWKWIHEPKGYHANFCLGPCPYIWSLDTQYSKVLALYNQHNPGASAAPCCVPQALEPLPIVYYVGRKPKVEQLSNMIVRSCKCSSpecifications
| TGFB1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_000660 | |
| TGFB1 | |
| TGFB1 (NP_000651, 279 a.a. ∼ 390 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7040 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| 4E1 | |
| Mouse monoclonal antibody raised against a partial recombinant TGFB1. | |
| TGFB1 | |
| CED/DPD1/TGFB/TGFbeta | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.