missing translation for 'onlineSavingsMsg'
Learn More
Learn More
transducin-like enhancer of split 6 (E(sp1) homolog, Drosophila), Mouse, Clone: 2E4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant TLE6.
Brand: Abnova H00079816-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MATRSSDWLRRPLGEDNQPETQLFWDKEPWFWHDTLTEQLWRIFAGVHDEKAKPRDRQQAPGLGQESKAPGSCDPGTDPCPEDASTPRPPEASSSPPEGSQDRNTSWGVVQEPPGRASRFLQSISWDPEDFEDAWKRPDALPGQSKRLAVPCKLEKMRILAHGELVLATAISSFTRHVFTCGRRGIKVWSLTGQVAEDRFPESHLPIQTPGAFLRTCLLSSNSRSLLTGGYNLASVSVWDLAAPSLHVKEQLPCAGLNCQALDANLDANLAFASFTSGVVRIWDLRDQSVVRDLKGYPDGVKSIVVKGYNIWTGGPDACLRCWDQRTIMKPLEYQFKSQIMSLSHSPQEDWVLLGMANGQQWLQSTSGSQRHMVGQKDSVILSVKFSPFGQWWASVGMDDFLGVYSMPAGTKVFEVPEMSPVTCCDVSSNNRLVVTGSGEHASVYQITYSpecifications
| transducin-like enhancer of split 6 (E(sp1) homolog, Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC020206 | |
| TLE6 | |
| TLE6 (AAH20206, 1 a.a. ∼ 449 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Signal Transduction | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2E4 | |
| Mouse monoclonal antibody raised against a full length recombinant TLE6. | |
| TLE6 | |
| FLJ14009/GRG6/MGC14966 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 79816 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction