Learn More
TNFRSF8, Mouse, Clone: 4C9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TNFRSF8.
Brand: Abnova H00000943-M02.100ug
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is expressed by activated, but not by resting, T and B cells. TRAF2 and TRAF5 can interact with this receptor, and mediate the signal transduction that leads to the activation of NF-kappaB. This receptor is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: QDRPFEDTCHGNPSHYYDKAVRRCCYRCPMGLFPTQQCPQRPTDCRKQCEPDYYLDEADRCTACVTCSRDDLVEKTPCAWNSSRVCECRPGMFCSTSAVNSCARCFFHSVCPASpecifications
| TNFRSF8 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001243 | |
| TNFRSF8 | |
| TNFRSF8 (NP_001234, 21 a.a. ∼ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 4C9 | |
| Mouse monoclonal antibody raised against a partial recombinant TNFRSF8. | |
| TNFRSF8 | |
| CD30/D1S166E/KI-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 943 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.