missing translation for 'onlineSavingsMsg'
Learn More

tumor protein p53 inducible protein 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16140896
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16140896 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16140896 Supplier Abnova Supplier No. H00009540B02P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human TP53I3 protein.

The protein encoded by this gene is similar to oxidoreductases, which are enzymes involved in cellular responses to oxidative stresses and irradiation. This gene is induced by the tumor suppressor p53 and is thought to be involved in p53-mediated cell death. It contains a p53 consensus binding site in its promoter region and a downstream pentanucleotide microsatellite sequence. P53 has been shown to transcriptionally activate this gene by interacting with the downstream pentanucleotide microsatellite sequence. The microsatellite is polymorphic, with a varying number of pentanucleotide repeats directly correlated with the extent of transcriptional activation by p53. It has been suggested that the microsatellite polymorphism may be associated with differential susceptibility to cancer. At least two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: MLAVHFDKPGGPENLYVKEVAKPSPGEGEVLLKVAASALNRADLMQRQGQYDPPPGASNILGLEASGHVAELGPGCQGHWKIGDTAMALLPGGGQAQYVTVPEGLLMPIPEGLTLTQAAAIPEAWLTAFQLLHLVGNVQAGDYVLIHAGLSGVGTAAIQLTRMAGAIPLVTAGSQKKLQMAEKLGAAAGFNYKKEDFSEATLKFTKGAGVNLILDCIGGSYWEKNVNCLALDGRWVLYGLMGGGDINGPLFSKLLFKRGSLITSLLRSRDNKYKQMLVNAFTEQILPHFSTEGPQRLLPVLDRIYPVTEIQEAHKYMEANKNIGKIVLELPQ

Specifications

Antigen tumor protein p53 inducible protein 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TP53I3 protein.
Formulation PBS with no preservative; pH 7.4
Gene TP53I3
Gene Accession No. NM_004881
Gene Alias PIG3
Gene Symbols TP53I3
Host Species Mouse
Immunogen TP53I3 (NP_004872, 1 a.a. ∼ 332 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9540
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.