missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tumor protein, translationally-controlled 1, Mouse, Clone: 2C4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TPT1.
Brand: Abnova H00007178-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: GVDIVMNHHLQETSFTKEAYKKYIKDYMKSIKGKLEEQRPERVKPFMTGAAEQIKHILANFKNYQFFIGENMNPDGMVALLDYREDGVTPYMIFFKDGLEMEKCSpecifications
| tumor protein, translationally-controlled 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC022436 | |
| TPT1 | |
| TPT1 (AAH22436, 35 a.a. ∼ 138 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Stem Cell Biology | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 2C4 | |
| Mouse monoclonal antibody raised against a partial recombinant TPT1. | |
| TPT1 | |
| FLJ27337/HRF/TCTP/p02 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 7178 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction