Learn More
tribbles homolog 3 (Drosophila), Mouse, Clone: 2G5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TRIB3.
Brand: Abnova H00057761-M06.100ug
Description
The protein encoded by this gene is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1. [provided by RefSeq
Sequence: PDRATAVATASRLGPYVLLEPEEGGRAYRALHCPTGTEYTCKVYPVQEALAVLEPYARLPPHKHVARPTEVLAGTQLLYAFFTRTHGDMHSpecifications
| tribbles homolog 3 (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC027484 | |
| TRIB3 | |
| TRIB3 (AAH27484, 56 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2G5 | |
| Mouse monoclonal antibody raised against a partial recombinant TRIB3. | |
| TRIB3 | |
| C20orf97/NIPK/SINK/SKIP3/TRB3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 57761 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Safety and Handling
- TRIB3 monoclonal antibody (M06), clone 2G5
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P233-Keep container tightly closed.
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P404-Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : sodiumphosphate dibasic, potassium phosphate monobasic, potassium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.