Learn More
tRNA splicing endonuclease 54 homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant TSEN54.
Brand: Abnova H00283989-A01.50uL
Description
This gene encodes a subunit of the tRNA splicing endonuclease complex, which catalyzes the removal of introns from precursor tRNAs. The complex is also implicated in pre-mRNA 3-prime end processing. Mutations in this gene result in pontocerebellar hypoplasia type 2
Sequence: LQTTHLPDGGVRLLEKSGGLEIIFDVYQADAVATFRKNNPGKPYARMCISGFDEPVPDLCSLKRLSYQSGDVPLIFALVDHGDISFYSFRDFTLPQDVGSpecifications
| tRNA splicing endonuclease 54 homolog (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant TSEN54. | |
| TSEN54 | |
| FLJ37147/PCH2A/SEN54L/sen54 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 283989 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_207346 | |
| TSEN54 | |
| TSEN54 (NP_997229, 427 a.a. ∼ 525 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.