missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tetraspanin 17, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TSPAN17 protein.
Brand: Abnova H00026262-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MPGKHQHFQEPEVGCCGKYFLFGFNIVFWVLGALFLAIGLWAWGEKGVLSNISALTDLGGLDPVWLFVVVGGVMSVLGFAGCIGALRENTFLLKFFSVFLGLIFFLELATGILAFVFKDWIRDQLNLFINNNVKAYRDDIDLQNLIDFAQEYWSCCGARGPNDWNLNIYFNCTDLNPSRERCGVPFSCCVRDPAEDVLNTQCGYDVRLKLELEQQGFIHTKGCVGQFEKWLQDNLIVVAGVFMGIALLQIFGICLAQNLVSDIKAVKANWSKWNDDFENHWLTPTISEVLSTAGPQQNSLTGAPGPAPPSRHVFFGLGGLYPEPTFKNWSpecifications
| tetraspanin 17 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human TSPAN17 protein. | |
| TSPAN17 | |
| FBX23/FBXO23/MGC14859/MGC71255/TM4SF17 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_130465.3 | |
| TSPAN17 | |
| TSPAN17 (NP_569732.2, 1 a.a. ∼ 329 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 26262 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction