Learn More
ubiquitin specific peptidase 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human USP1 protein.
Brand: Abnova H00007398-B01P.50ug
Description
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. The protein specifically deubiquitinates a protein in the Fanconi anemia (FA) DNA repair pathway. Alternate transcriptional splice variants have been characterized. [provided by RefSeq
Sequence: MPGVIPSESNGLSRGSPSKKNRLSLKFFQKKETKRALDFTDSQENEEKASEYRASEIDQVVPAAQSSPINCEKRENLLPFVGLNNLGNTCYLNSILQVLYFCPGFKSGVKHLFNIISRKKEALKDEANQKDKGNCKEDSLASYELICSLQSLIISVEQLQASFLLNPEKYTDELATQPRRLLNTLRELNPMYEGYLQHDAQEVLQCILGNIQETCQLLKKEEVKNVAELPTKVEEIPHPKEEMNGINSIEMDSMRHSEDFKEKLPKGNGKRKSDTEFGNMKKKVKLSKEHQSLEENQRQTRSKRKATSDTLESPPKIIPKYISENESPRPSQKKSRVKINWLKSATKQPSILSKFCSLGKITTNQGVKGQSKENECDPEEDLGKCESDNTTNGCGLESPGNTVTPVNVNEVKPINKGEEQIGFELVEKLFQGQLVLRTRCLECESLTERREDFQDISVPVQEDELSKVEESSEISPEPKTEMKTLRWAISQFASVERIVGEDKYFCENCHHYTEAERSLLFDKMPEVITIHLKCFAASGLEFDCYGGGLSKINTPLLTPLKLSLEEWSTKPTNDSYGLFAVVMHSGITISSGHYTASVKVTDLNSLELDKGNFVVDQMCEIGKPEPLNEEEARGVVENYNDEEVSIRVGGNTQPSKVLNKKNVEAIGLLGGQKSKADYELYNKASNPDKVASTAFAENRNSETSDTTGTHESDRNKESSDQTGINISGFENKISYVVQSLKEYEGKWLLFDDSEVKVTEEKDFLNSLSPSTSPTSTPYLLFYKKLSpecifications
| ubiquitin specific peptidase 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human USP1 protein. | |
| USP1 | |
| UBP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_003368.4 | |
| USP1 | |
| USP1 (NP_003359.3, 1 a.a. ∼ 785 a.a) full-length human protein. | |
| 50 μg | |
| Ubiquitin/Proteasome | |
| 7398 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.