missing translation for 'onlineSavingsMsg'
Learn More

Vβ 12 TCR Mouse anti-Mouse, FITC, Clone: MR11-1, BD

Product Code. 15807138
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15807138 0.25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15807138 Supplier BD Biosciences Supplier No. 553300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The MR11-1 antibody reacts with the Vβ 12 T-cell Receptor (TCR) of mice having the b haplotype (e.g., C57BL, C58, DBA/1) of the Tcrb gene complex1 The Tcrb-V12 gene locus is deleted in mice having the a (e.g., C57BR, C57L, SJL, SWR) or c (e.g., RIII)

Host Species: Mouse
Clone: MR11-1
Isotype: IgG1 κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 12 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone MR11-1
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Not Reported
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.