Learn More
Vα 3.2[b,c] TCR Rat anti-Mouse, FITC, Clone: RR3-16, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553219
Beschreibung
The RR3-16 antibody reacts with the Vα 3.2 T-Cell Receptor (TCR) of mice having the b (e.g., C57BL) and c (e.g., SWR, SJL, NZB, NOD) haplotypes of the Tcra gene complex, but not with TCR encoded by other members of Tcra-V3 gene subfamily. RR3-16 ant
Host Species: Rat
Clone: RR3-16
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Cytolytic T-Cell Clone OH6
Flow Cytometry
Spezifikation
| Vα 3.2[b,c] TCR | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse Cytolytic T-Cell Clone OH6 | |
| 0.25 mg | |
| Immunology | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| RR3-16 | |
| FITC | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.