Learn More
Vβ 6 TCR Rat anti-Mouse, Biotin, Clone: RR4-7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553192
Description
The RR4-7 antibody reacts with the Vβ 6 T-Cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL) and b (e.g., A, BALB/c, CBA/Ca, C3H/He, C57BL, DBA/1) haplotypes of the Tcrb gene complex. The Tcrb-V6 gene locus is deleted in mice having th
Host Species: Rat
Clone: RR4-7
Isotype: IgG2b λ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Helper T-Cell Clone OI11
Flow Cytometry
Specifications
| Vβ 6 TCR | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| C57BL/6 Mouse Helper T-Cell Clone OI11 | |
| 0.25 mg | |
| Immunology | |
| Mouse | |
| IgG2b λ |
| Flow Cytometry | |
| RR4-7 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.