missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vasopressin, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Vasopressin [Arg8].
Brand: Abnova PAB5189.150ug
This item is not returnable.
View return policy
Description
Sequence: CYFQNCPRGSpecifications
| Vasopressin | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Vasopressin [Arg8]. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| A synthetic peptide (conjugated with KLH) corresponding to vasopressin [Arg8]. | |
| RUO | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction