Learn More
Viperin Mouse, PE, Clone: MaP.VIP, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565196
Description
The MaP.VIP monoclonal antibody specifically binds to Virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible (Viperin), which is also known as Radical S-adenosyl methionine domain-containing protein 2 (Rsad2). Viperin is a 43 kDa protein that is localized to the endoplasmic reticulum and lipid droplets. It functions in protective responses to a number of different DNA and RNA viruses. Viperin expression is induced in cells by different agents such as, Type I interferons, Interferon- γ(IFN- γ), DNA and RNA viral proteins, Polyinosinic:polycytidylic acid, and lipopolysaccharide. Various cell types can be induced to express Viperin including T and B lymphocytes, macrophages, granulocytes, dendritic cells, fibroblasts, endothelial cells, and epithelial cells.
Specifications
| Viperin | |
| Monoclonal | |
| 0.2mg/mL | |
| Rsad2; Vig1; cig5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Flow Cytometry | |
| MaP.VIP | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide | |
| Mouse Viperin Recombinant Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.