Learn More
VNN1, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00008876-PW1.1Set
Product Code. 16172924
  • € 843.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-VNN1 (300μL), Antibody pair for WB: mouse purified polyclonal anti-VNN1 (50μg)
Specifications
VNN1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
HDLCQ8/MGC116930/MGC116931/MGC116932/MGC116933/Tiff66
Mouse, Rabbit
RUO
8876
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
VNN1
VNN1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-VNN1 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-VNN1 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.