Learn More
WD repeat domain 36, Mouse, Clone: 1D6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant WDR36.
Brand: Abnova H00134430-M01.100ug
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Mutations in this gene have been associated with adult-onset primary open-angle glaucoma (POAG). [provided by RefSeq
Sequence: SGIETELRSLSPDCGGSIEVMQSFLKMIGMMLDRKRDFELAQAYLALFLKLHLKMLPSEPVLLEEITNLSSQVEENWTHLQSLFNQSMCILNYLKSALLSpecifications
| WD repeat domain 36 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_139281 | |
| WDR36 | |
| WDR36 (NP_644810, 853 a.a. ∼ 951 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 134430 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1D6 | |
| Mouse monoclonal antibody raised against a partial recombinant WDR36. | |
| WDR36 | |
| DKFZp686I1650/GLC1G/TA-WDRP/TAWDRP/UTP21 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.