Learn More
WEE1 homolog (S. pombe), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human WEE1 protein.
Brand: Abnova H00007465-D01P.100ug
Description
This gene encodes a nuclear protein, which is a tyrosine kinase belonging to the Ser/Thr family of protein kinases. This protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase. [provided by RefSeq
Sequence: MSFLSRQQPPPPRRAGAACTLRQKLIFSPCSDCEEEEEEEEEEGSGHSTGEDSAFQEPDSPLPPARSPTEPGPERRRSPGPAPGSPGELEEDLLLPGACPGADEAGGGAEGDSWEEEGFGSSSPVKSPAAPYFLGSSFSPVRCGGPGDASPRGCGARRAGEGRRSPRPDHPGTPPHKTFRKLRLFDTPHTPKSLLSKARGIDSSSVKLRGSSLFMDTEKSGKREFDVRQTPQVNINPFTPDSLLLHSSGQCRRRKRTYWNDSCGEDMEASDYELEDETRPAKRITITESNMKSRYTTEFHELEKIGSGEFGSVFKCVKRLDGCIYAIKRSKKPLAGSVDEQNALREVYAHAVLGQHSHVVRYFSAWAEDDHMLIQNEYRNGGSLADAISENYRIMSYFKEAELKDLLLQVGRGLRYIHSMSLVHMDIKPSNIFISRTSIPNAASEEGDEDDWASNKVMFKIGDLGHVTRISSPQVEEGDSRFLANEVLQENYTHLPKADIFALALTVVCAAGAEPLPRNGDQWHEIRQGRLPRIPQVLSQEFTELLKVMIHPDPERRPSAMALVKHSVLLSASRKSAEQLRIELNAEKFKNSLLQKELKKAQMAKAAAEERALFTDRMATRSTTQSNRTSRLIGKKMNRSVSLTIYSpecifications
| WEE1 homolog (S. pombe) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human WEE1 protein. | |
| WEE1 | |
| DKFZp686I18166/FLJ16446/WEE1A/WEE1hu | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| In situ PLA, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC051831.1 | |
| WEE1 | |
| WEE1 (AAH51831.1, 1 a.a. ∼ 646 a.a) full-length human protein. | |
| 100 μg | |
| Cell Cycle | |
| 7465 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.