missing translation for 'onlineSavingsMsg'
Learn More

XRCC4 Mouse anti-Human, Unlabeled, Clone: 4, BD

Product Code. 15835959
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835959 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15835959 Supplier BD Biosciences Supplier No. 611506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DNA double-strand breaks (DSB) are generated during intrinsic eukaryotic DNA recombination events such as assembly of antigen receptor genes and meiotic and mitotic recombination. DSB repair proteins are also required to repair breaks induced by extrinsic factors such as ionizing radiation and mutagenic chemicals. DNA-PKcs, Ku70/Ku80, DNA ligase IV, and X-Ray Cross Complementation group 4 (XRCC4) are DSB proteins involved in both V(D)J recombination and DNA double-stranded break repair. XRCC4 activates DNA ligase IV and cells deficient in XRCC4 inefficiently form coding joints and signal joints during V(D)J recombination. XRCC4 contains a C-terminal nuclear localization sequence (NLS) and multiple phosphorylation sites, binds DNA, and is an effective substrate for DNA-PK. Phosphorylation of XRCC4 has no effect on its interactions with DNA ligase IV or end-joining activity, but can inhibit its DNA binding activity. Mice deficient in XRCC4 exhibit defects in lymphogenesis and apoptotic death of postmitotic neurons during neurogenesis. Thus, XRCC4 is a ubiquitous protein involved in DNA end joining during DNA recombination and repair, which is critical for cell growth and survival.

Host Species: Mouse
Clone: 4
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human XRCC4 aa. 53-168
Formula Weight [Chemical]: 55kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen XRCC4
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Description X-Ray Cross Complementation group 4
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human XRCC4 aa. 53-168
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.