missing translation for 'onlineSavingsMsg'
Learn More
Learn More
zinc finger and BTB domain containing 25, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZBTB25 protein.
Brand: Abnova H00007597-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MDTASHSLVLLQQLNMQREFGFLCDCTVAIGDVYFKAHRAVLAAFSNYFKMIFIHQTSECIKIQPTDIQPDIFSYLLHIMYTGKGPKQIVDHSRLEEGIRFLHADYLSHIATEMNQVFSPETVQSSNLYGIQISTTQKTVVKQGLEVKEAPSSNSGNRAAVQGDHPQLQLSLAIGLDDGTADQQRACPATQALEEHQKPPVSIKQERCDPESVISQSHPSPSSEVTGPTFTENSVKIHLCHYCGERFDSRSNLRQHLHTHVSGSLPFGVPASILESNDLGEVHPLNENSEALECRRLSSFIVKENEQQPDHTNRGTTEPLQISQVSLISKDTEPVELNCNFSFSRKRKMSCTICGHKFPRKSQLLEHMYTHKGKSYRYNRCQRFGNALAQRFQPYCDSWSDVSLKSSRLSQEHLDLPCALESELTQENVDTILVESpecifications
| zinc finger and BTB domain containing 25 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ZBTB25 protein. | |
| ZBTB25 | |
| KUP/ZNF46 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_006977.2 | |
| ZBTB25 | |
| ZBTB25 (NP_008908.2, 1 a.a. ∼ 435 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 7597 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction