Learn More
anti-ZNF350, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00059348-PW1.1Set
Product Code. 16133604
  • € 843.00 / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-ZNF350 (300μg), Antibody pair for WB: rabbit polyclonal anti-ZNF350 (50μL)
Specifications
ZNF350
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
ZBRK1/ZFQR
Mouse, Rabbit
RUO
59348
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
ZNF350
ZNF350
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-ZNF350 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-ZNF350 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.