missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ubiquitin protein ligase E3 component n-recognin 3 (putative) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ZNF650 protein.
Brand: Abnova H00130507-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MNKRIIEEICRKVTPPVPPKKVTAAEKKTLDKEERRQKARERQQKLLAEFASRQKSFMETAMDVDSPENDIPMEITTAEPQVSEAVYDCVICGQSGPSSEDRPTGLVVLLQASSVLGQCRDNVEPKKLPISEEEQIYPWDTCAAVHDVRLSLLQRYFKDSSCLLAVSIGWEGGVYVQTCGHTLHIDCHKSYMESLRNDQVLQGFSVDKGEFTCPLCRQFANSVLPCYPGSNVENNPWQRPSNKSIQDLIKEVEELQGRPGAFPSETNLSKEMESVMKDIKNTTQKKYRDYSKTPGSPDNDFLFMYSVARTNLELELIHRGGNLCSGGASTAGKRSCLNQLFHVLALHMRLYSIDSEYNPWRKLTQLEEMNPQLGYEEQQPEVPILYHDVTSLLLIQILMMPQPLRKDHFTCIVKVLFTLLYTQALAALSVKCSEEDRSAWKHAGALKKSTCDAEKSYEVLLSFVISELFKGKLYHEEGTQECAMVNPIAWSPESMEKCLQDFCLPFLRITSLLQHHLFGEDLPSCQEEEEFSVLASCLGLLPTFYQTEHPFISASCLDWPVPAFDIITQWCFEIKSFTERHAEQGKALLIQESKWKLPHLLQLPENYNTIFQYYHRKTCSVCTKVPKDPAVCLVCGTFVCLKGLCCKQQSYCECVLHSQNCGAGTGIFLLINASVIIIIRGHRFCLWGSVYLDAHGEEDRDLRRGKPLYICKERYKVLEQQWISHTFDHINKRWGPHYNGLSpecifications
| ubiquitin protein ligase E3 component n-recognin 3 (putative) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ZNF650 protein. | |
| UBR3 | |
| DKFZp434P117/DKFZp686N10185/FLJ37422/KIAA2024/MGC126489/ZNF650 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_172070.2 | |
| UBR3 | |
| ZNF650 (NP_742067.2, 1 a.a. ∼ 741 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 130507 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction