Learn More
Cole-Parmer™ Hexagonal Polystyrene Weighing Dishes
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
20 mL
50 mL
200 mL
350 mL
Unit Size:
Pack of 500
Description
- Translucent anti-static, polystyrene
- Resistant to most dilute acids, aqueous solutions and alcohol bases
- General purpose sample holders
- Unique hexagonal design for non-slip handling and pouring
- Disposable
- Biologically inert
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 350 mL |
| Disposable | Single-use |
| Material | Polystyrene |
| Shape | Hexagonal |
| Type | Antistatic Weighing Dish |
| For Use With (Application) | For weighing, dispensing, or pouring liquids or solids |
| Size | X-Large |
| Depth (Metric) | 14 cm |
| Diameter (English) | 5.5 in. |
| Show More |