missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANTXR2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00118429-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MVAERSPARSPGSWLFPGLWLLVLSGPGGLLRAQEQPSCRRAFDLYFVLDKSGSVANNWIEIYNFVQQLAERFVSPEMRLSFIVFSSQATIILPLTGDRGKISKGLEDLKRVSPVGETYIHEGLKLANEQIQKAGGLKTSSIIIALTDGKLDGLVPSYAEKEAKISRSLGASVYCVGVLDFEQAQLERIADSKEQVFPVKGGFQALKGIINSILAQSCTEILELQPSSVCVGEEFQIVLSGRGFMLGSRNGSVLCTYTVNETYTTSVKPVSVQLNSMLCPAPILNKAGETLDVSVSFNGGKSVISGSLIVTATECSNGIAAIIVILVLLLLLGIGLMWWFWPLCCKVVIKDPPPPPAPAPKEEEEEPLPTKKWPTVDASYYGGRGVGGIKRMEVRWGDKGSTEEGARLEKAKNAVVKIPEETEEPIRPRPPRPKPTHQPPQTKWYTPIKGRLDALWALLRRQYDRVSLMRPQEGDEGRCINFSRVPSQSpecifications
| ANTXR2 | |
| Polyclonal | |
| No additive | |
| NM_058172.3 | |
| ANTXR2 | |
| ANTXR2 (NP_477520.2, 1 a.a. ∼ 488 a.a) full-length human protein. | |
| RUO | |
| 118429 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| anthrax toxin receptor 2 | |
| CMG-2/CMG2/FLJ31074/ISH/JHF/MGC111533/MGC45856 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction