missing translation for 'onlineSavingsMsg'
Learn More

ANXA2R Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16862465 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16862465 20 μL 20µL
16852465 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16862465 Supplier Proteintech Supplier No. 238381AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C5orf39 may act as a receptor for annexin II on marrow stromal cells to induce osteoclast formation.

Specifications

Antigen ANXA2R
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANXA2R
Gene Accession No. Q3ZCQ2
Gene Alias Annexin 2 receptor, Annexin II receptor, AX2R, AXIIR, C5orf39
Gene Symbols ANXA2R
Host Species Rabbit
Immunogen ANXA2R Fusion Protein Ag20807
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389289
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.