Learn More
Invitrogen™ Anza™ 18 PacI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN0186
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15555700
  • € 382.00 / 1 set
Estimated Shipment: 14-05-2024
to see stock.
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Specifications
Specifications
500 μL Anza 18 PacI
1 mL Anza Buffer (10X)
1 mL Anza Red Buffer (10X)
Store at -20°C.
Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
1000 U
Traditional Cloning
Approved for shipment on Wet or Dry Ice
Pac I
Yes
No
Restriction Enzyme Cloning System
Anza™