missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOC3 Monoclonal Antibody (174-5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548984
This item is not returnable.
View return policy
Description
The monoclonal antibody 174-5 has been shown to cross-react with rat VAP-1 and to inhibit lymphocyte infiltration in rat liver allograft rejection.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Specifications
| AOC3 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and no preservative | |
| Q16853 | |
| AOC3 | |
| Purified vessels from human peripheral lymph nodes. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunocytochemistry | |
| 174-5 | |
| Unconjugated | |
| AOC3 | |
| Amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8639 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction