missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOC3 Recombinant Mouse Monoclonal Antibody (TK8-14)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551832
This item is not returnable.
View return policy
Description
This antibody is specific for human VAP-1, a glycosylated homodimeric membrane protein consisting of two 90 kDa subunits connected by disulfide bonds. The recognized epitope is present both in dimeric and monomeric forms of VAP-1.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
Specifications
| AOC3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q16853 | |
| AOC3 | |
| Affinity purified VAP-1 from tonsil stroma of human origin. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| TK8-14 | |
| Unconjugated | |
| AOC3 | |
| amine oxidase, copper containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Aoc3; Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; semicarbazide-sensitive amine oxidase; SSAO; VAP1; VAP-1; Vascular adhesion protein 1; VP97 | |
| Mouse | |
| Protein A | |
| RUO | |
| 8639 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction