missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOF1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81895AF647
This item is not returnable.
View return policy
Description
AOF1 Polyclonal antibody specifically detects AOF1 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| AOF1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 221656 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| AOF1, bA204B7.3, C6orf193, dJ298J15.2, DKFZp686I0412, flavin containing 1, flavin-containing amine oxidase domain-containing protein 1, FLJ33898, FLJ34109, FLJ43328, LSD2, lysine (K)-specific demethylase 1B, lysine-specific histone demethylase 1B, lysine-specific histone demethylase 2 | |
| AOF1 antibody was raised against a 17 amino acid peptide near the amino terminus of human AOF1. The immunogen is located within amino acids 80 - 130 of AOF1. Amino Acid Squence: FDHYYRSHKDGYDKYT | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction