missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AOF2 Mouse anti-Human, Clone:3F9D2, Abnova™
Mouse monoclonal antibody raised against recombinant human AOF2.
Brand: Abnova MAB17847.100ug
This item is not returnable.
View return policy
Description
This gene encodes a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain. This protein is a component of several histone deacetylase complexes, though it silences genes by functioning as a histone demethylase. Alternative splicing results in multiple transcript variants. [provided by RefSeq]Specifications
| AOF2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-400) Immunofluorescence (1:200-1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user. |
|
| AOF2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3F9D2 | |
| amine oxidase (flavin containing) domain 2 | |
| In PBS (0.05% sodium azide) | |
| BHC110, KDM1, KIAA0601, LSD1 | |
| Recombinant protein corresponding to amino acids 55-263 of human AOF2 from E. coli. | |
| 23028 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction