missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP-2 beta/TFAP2B Antibody (6F7G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61858
This item is not returnable.
View return policy
Description
AP-2 beta/TFAP2B Monoclonal specifically detects AP-2 beta/TFAP2B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| AP-2 beta/TFAP2B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| activating enhancer binding protein 2 beta, Activating enhancer-binding protein 2-beta, AP-2B, AP2-B, AP2-beta, CHAR, MGC21381, transcription factor AP-2 beta (activating enhancer binding protein 2 beta), transcription factor AP-2 beta (activating enhancer-binding protein 2 beta), transcription factor AP-2-beta | |
| Mouse | |
| 50.5 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 6F7G1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TFAP2B | |
| Purified recombinant fragment of human TFAP2B (AA: 84-193) expressed in E. Coli. | |
| Protein G purified | |
| Neuroscience | |
| 7021 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction