missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP-2 gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98897-50ul
This item is not returnable.
View return policy
Description
AP-2 gamma Polyclonal antibody specifically detects AP-2 gamma in Human samples. It is validated for Western Blot
Specifications
| AP-2 gamma | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Activating enhancer-binding protein 2 gamma, AP2-GAMMA, ERF1, estrogen receptor factor 1, hAP-2g, TFAP2G, transcription factor AP-2 gamma, transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma), transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma), Transcription factor ERF-1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human AP-2 gamma. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7022 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction