missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP repeat Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119810
This item is not returnable.
View return policy
Description
Positive Control: Recombinant protein lysates.
The C9orf72 GGGGCC repeat sequence codes five repeat peptide GA repeat; GAGAGAGAGA, GP repeat; GPGPGPGPG, GR repeat; GRGRGRGRG, AP repeat; APAPAPAPA and PR repeat; PRPRPRPRP. It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.
Specifications
| AP repeat | |
| Polyclonal | |
| Unconjugated | |
| AP repeat; APAPAPAPA; C9RAN; GGGCC; poly AP | |
| Synthetic peptide immune sequence is CAPAPAPAPA. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction