missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP1AR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566133
This item is not returnable.
View return policy
Description
Immunogen sequence: RIVQQYHPSNN GEYQSSGPED DFESCLRNMK SQYEVFRSSR LSSDATVLTP NTESSCDLMT KTKSTSGNDD STSLDLEWED Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
AP1AR is necessary for AP-1 dependent transport between the trans-Golgi network and endosomes. It regulates the membrane association of AP1G1/Gamma1-adaptin, one of the subunits of the AP-1 adapter complex. The direct interaction with AP1G1/Gamma1-adaptin attenuates the release of the AP-1 complex from membranes.
Specifications
| AP1AR | |
| Polyclonal | |
| Unconjugated | |
| AP1AR | |
| 2c18; AA407621; adapter-related protein complex 1-associated regulatory protein; adaptor related protein complex 1 associated regulatory protein; adaptor-related protein complex 1 associated regulatory protein; adaptor-related protein complex 1-associated regulatory protein; AP-1 complex-associated regulatory protein; AP-1 complex-associated regulatory protein-like; AP1AR; C4orf16; C78909; Gadkin; gamma-1-adaptin brefeldin A resistance protein; gamma1-adaptin brefeldin A resistance protein, gamma-BAR; Gamma-A1-adaptin and kinesin interactor; gamma-BAR; GBAR; PRO0971; RGD1311435 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55435 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q63HQ0 | |
| AP1AR | |
| Recombinant Human AP1AR. Recombinant protein control fragment (Product #RP-106817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction