missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (3B5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-74359
This item is not returnable.
View return policy
Description
AP2 alpha Monoclonal specifically detects AP2 alpha in Human, Mouse, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| AP2 alpha | |
| Monoclonal | |
| Unconjugated | |
| P05549 | |
| TFAP2A | |
| N-terminus of human AP2a. | |
| Protein G purified | |
| RUO | |
| Primary | |
| AP-2 (3B5) | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3B5 | |
| Western Blot 1:50 - 1:200, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:100 - 1:250, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:50 | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| Mouse | |
| 48 kDa | |
| 100 μL | |
| Cancer, Transcription Factors and Regulators | |
| 7020 | |
| Human, Mouse, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction