missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (AP2a 8G8/5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2639-0.025mg
This item is not returnable.
View return policy
Description
AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry.
Specifications
| AP2 alpha | |
| Monoclonal | |
| 0.82 mg/ml | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| TFAP2A | |
| Bacterialy expressed AP-2a/AP-2g fusion protein. | |
| Protein A purified | |
| RUO | |
| Primary | |
| AP2a 8G8/5 is specific for the alpha form of AP-2.Antigen recovery using microwave or pressure cooking required for paraffin embedded tissue. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| AP2a 8G8/5 | |
| Unconjugated | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| Mouse | |
| 50 kDa | |
| 0.025 mg | |
| Cancer, Transcription Factors and Regulators | |
| 7020 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction