Learn More
Invitrogen™ AP2 alpha Monoclonal Antibody (R.387.0)
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  Invitrogen™ MA514856
Product Code. 11584821
  • € 520.00 / 100µL
Description
It is not recommended to aliquot this antibody.
TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AP2 alpha
Monoclonal
94.8 μg/mL
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
P05549, P34056, P58197
TFAP2A
Synthetic peptide corresponding to human AP-2a.
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
Western Blot, Immunocytochemistry
R.387.0
Unconjugated
TFAP2A
activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2
Rabbit
Affinity Chromatography
RUO
21418, 306862, 7020
-20°C