missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2B1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | AP2B1 |
|---|---|
| Clone | 1A3B6 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236150
|
Proteintech
68349-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233731
|
Proteintech
68349-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AP2B1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AP2B1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The AP2B1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| AP2B1 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 163 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1A3B6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Rabbit, Pig | |
| ADTB2; AP105B; AP2-BETA; CLAPB1; DKFZp781K0743 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title