missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2M1 Antibody (4F1B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61866-0.025ml
This item is not returnable.
View return policy
Description
AP2M1 Monoclonal specifically detects AP2M1 in Human, Monkey samples. It is validated for Western Blot, ELISA.
Specifications
| AP2M1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Adaptin-mu2, adaptor-related protein complex 2, mu 1 subunit, AP-2 mu chain, Clathrin assembly protein complex 2 medium chain, clathrin coat adaptor protein AP50, Clathrin coat assembly protein AP50, Clathrin coat-associated protein AP50, clathrin-associated/assembly/adaptor protein, medium 1, HA2 50 kDa subunit, KIAA0109, mu subunit, Plasma membrane adaptor AP-2 50 kDa protein, plasma membrane adaptor AP-2 50kDA protein | |
| Mouse | |
| 49.7 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, ELISA | |
| 4F1B2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AP2M1 | |
| Purified recombinant fragment of human AP2M1 (AA: 298-435) expressed in E. Coli. | |
| Protein G purified | |
| Phospho Specific, Signal Transduction | |
| 1173 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction