missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3D1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92046-0.02ml
This item is not returnable.
View return policy
Description
AP3D1 Polyclonal antibody specifically detects AP3D1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| AP3D1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Adapter-related protein complex 3 subunit delta-1, adaptin, delta, adaptor-related protein complex 3, delta 1 subunit, ADTD, AP-3 complex delta subunit, partial CDS, AP-3 complex subunit delta, AP-3 complex subunit delta-1, delta adaptin, Delta-adaptin, delta-adaptin, partial CDS, hBLVR, subunit of putative vesicle coat adaptor complex AP-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 520-700 of human AP3D1 (NP_003929.4). AVYVQNVVKLYASILQQKEQAGEAEGAQAVTQLMVDRLPQFVQSADLEVQERASCILQLVKHIQKLQAKDVPVAEEVSALFAGELNPVAPKAQKKVPVPEGLDLDAWINEPLSDSESEDERPRAVFHEEEQRRPKHRPSEADEEELARRREARKQEQANNPFYIKSSPSPQKRYQDTPGVE | |
| 0.02 mL | |
| Signal Transduction | |
| 8943 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur