missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3D1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92046-0.1ml
This item is not returnable.
View return policy
Description
AP3D1 Polyclonal antibody specifically detects AP3D1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| AP3D1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Adapter-related protein complex 3 subunit delta-1, adaptin, delta, adaptor-related protein complex 3, delta 1 subunit, ADTD, AP-3 complex delta subunit, partial CDS, AP-3 complex subunit delta, AP-3 complex subunit delta-1, delta adaptin, Delta-adaptin, delta-adaptin, partial CDS, hBLVR, subunit of putative vesicle coat adaptor complex AP-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 520-700 of human AP3D1 (NP_003929.4). AVYVQNVVKLYASILQQKEQAGEAEGAQAVTQLMVDRLPQFVQSADLEVQERASCILQLVKHIQKLQAKDVPVAEEVSALFAGELNPVAPKAQKKVPVPEGLDLDAWINEPLSDSESEDERPRAVFHEEEQRRPKHRPSEADEEELARRREARKQEQANNPFYIKSSPSPQKRYQDTPGVE | |
| 0.1 mL | |
| Signal Transduction | |
| 8943 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction