missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3M1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27068SS
This item is not returnable.
View return policy
Description
AP3M1 Polyclonal specifically detects AP3M1 in Human, Bovine, Canine, Chicken, Chinese Hamster, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AP3M1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| adaptor-related protein complex 3, mu 1 subunit, AP-3 adapter complex mu3A subunit, MGC22164, mu-3A subunit, mu3A-adaptin | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 26985 | |
| Human, Bovine, Canine, Chicken, Chinese Hamster, Equine, Other | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml | |
| Q9Y2T2 | |
| AP3M1 | |
| The immunogen for this antibody was a portion of amino acid 140-190 of human AP3M1. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen for the AP3 antibody is 100% homologous in human, rhesus monkey, wild pig, dog, cow, horse, opossum, orangutan, mouse, rat, wild turkey, zebrafinch, chicken, new world monkey, Chinese hamster, macaque, rabbit, Carolina anole and platypus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction