missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP4M1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119950
This item is not returnable.
View return policy
Description
Positive Control: A. thaliana tissue. Subcellular Location: Trans-Golgi network, coated pit.
This gene encodes a subunit of the heterotetrameric AP-4 complex. The encoded protein belongs to the adaptor complexes medium subunits family. This AP-4 complex is involved in the recognition and sorting of cargo proteins with tyrosine-based motifs from the trans-Golgi network to the endosomal-lysosomal system.
Specifications
| AP4M1 | |
| Polyclonal | |
| Unconjugated | |
| 4930443L05Rik; adapter-related protein complex 4 mu-1 subunit; adapter-related protein complex 4 subunit mu-1; adaptor related protein complex 4 mu 1 subunit; adaptor related protein complex 4, mu 1 subunit; adaptor-related protein complex 4 subunit mu-1; adaptor-related protein complex 4, mu 1 subunit; adaptor-related protein complex AP-4 mu4 subunit; adaptor-related protein complex AP-4, mu 1; AP-4 adapter complex mu subunit; AP-4 adaptor complex mu subunit; AP-4 complex subunit mu-1; Ap4m1; ap4m1 {ECO:0000312; Ap4m4; CPSQ3; Mu subunit of AP-4; mu4; MU-4; mu4-adaptin; Mu-adaptin-related protein 2; mu-adaptin-related protein-2; MUARP2; mu-ARP2; RGD:1310233}; SPG50 | |
| Synthetic peptide within A. thaliana AP-4 complex subunit mu aa 380-430/451. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction