missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APAF-1 Antibody (2E12), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80092IR
This item is not returnable.
View return policy
Description
APAF-1 Monoclonal antibody specifically detects APAF-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| APAF-1 | |
| Monoclonal | |
| DyLight 755 | |
| APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
| Recombinant human APAF-1 (aa 1-464) containing the N-terminal CARD and CED-4 homologous domains. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
| 317 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2000000000000 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction