missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APBA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17217623 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17217623 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17217623 Supplier Invitrogen™ Supplier No. PA5114856

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total APBA1.

The munc-18 interacting protein (Mint) protein family is a group of evolutionarily conserved adaptor proteins that function in membrane transport and organization. In mammals, there exist three Mint isoforms, Mint1, 2, and 3. Although there is little amino acid sequence conservation in the amino-terminal half, the carboxy-terminal half of these proteins is highly conserved. Within this conserved portion there exists a phosphotyrosine-binding (PTB) and a PSD-95/DLG-A/ZO-1 (PDZ) domain, which function as protein interaction modules. Mint1 and 2 appear to be expressed exclusively in the brain and are found to bind to Munc18, an essential component of the synaptic vesicle fusion machinery. Mint3 is ubiquitously expressed in all tissues and is expressed at the lowest levels in the brain and testis. Studies show that mint3 does not interact with munc-18. Mint3 has been found to interact with the Alzheimer's Disease-related amyloid precursor protein (APP) and does so through its PTB and PDZ domains. It has been suggested that mint3 links APP to other transport machinery components, thereby regulating it transport, endocytosis, and metabolism. Abnormal APP metabolism has been shown to be the cause of an early-onset type of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APBA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene APBA1
Gene Accession No. B2RUJ5, O35430, Q02410
Gene Alias 6430513E09Rik; adapter protein X11alpha; adaptor protein X11alpha; amyloid beta (A4) precursor protein binding, family A, member 1; amyloid beta (A4) precursor protein-binding family A APBA1: amyloid beta (A4) precursor protein-binding family A member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, APBA1: amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, member 1; amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta A4 precursor protein-binding family A member 1; amyloid beta precursor protein binding family A member 1; Amyloid-beta A4 precursor protein-binding family A member 1; APBA1; D9S411E; LIN10; Lin-10; Mint; MINT1; Mint-1; neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; Urop11; UROP11-110; X11; X11A; X11ALPHA
Gene Symbols APBA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human APBA1(Accession Q02410), corresponding to amino acid residues M1-H31.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319924, 320, 83589
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.