Learn More
APBB1IP Mouse anti-Human, Clone: OTI1F2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809154
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.Specifications
| APBB1IP | |
| Monoclonal | |
| Unconjugated | |
| APBB1IP | |
| INAG1, PREL1, RARP1, RIAM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54518 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1F2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q7Z5R6 | |
| APBB1IP | |
| Full length human recombit protein of human APBB1IP produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.