missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APBB1IP Monoclonal Antibody (OTI5B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809244
This item is not returnable.
View return policy
Description
RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.Specifications
| APBB1IP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z5R6 | |
| APBB1IP | |
| Full length human recombit protein of human APBB1IP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI5B3 | |
| Unconjugated | |
| APBB1IP | |
| INAG1, PREL1, RARP1, RIAM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54518 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction