missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APBB2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15782741
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15782741 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782741 Supplier Invitrogen™ Supplier No. PA554816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SEKLEGKEPH PQDSSSCEIL PSQPRRTKSF LNYYADLETS ARELEQNRGN HHGTAEEKSQ PVQGQASTII GNGDLLLQKP NRPQSSPEDG QVATVSSSPE TKKDHPKTGA KTDCALHRIQ NLAPS Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 23%.

The protein encoded by this gene interacts with the cytoplasmic domains of amyloid beta precursor protein and amyloid beta precursor-like protein 2. This protein contains two phosphotyrosine binding domains, which are thought to function in signal transduction. Polymorphisms in this gene have been associated with Alzheimer's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APBB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Apbb2
Gene Accession No. Q92870
Gene Alias 2310007D03Rik; amyloid beta (A4) precursor protein-binding, family B, member 2; amyloid beta (A4) precursor protein-binding, family B, member 2 (Fe65-like); amyloid beta A4 precursor protein-binding family B member 2; amyloid beta A4 precursor protein-binding, family B, member 2; amyloid beta precursor protein binding family B member 2; amyloid beta precursor protein-binding family B member 2; amyloid-beta A4 precursor protein-binding family B member 2; Apbb2; FE65L; FE65L1; Fe65-like 1; Protein Fe65-like 1; RGD1562438; Rirl1; TR2L; Zfra; zinc finger-like protein
Gene Symbols Apbb2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APBB2. Recombinant protein control fragment (Product #RP-102098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.